GAK(cyclin-G-associated kinase) belongs to the protein kinase superfamily. GAK plays a more positive role as a modulator of signal transduction, by relaying the phosphorylation signal from an unidentified tyrosine kinase to unidentified GAK phosphorylation targets. It may be acting as an auxilin homolog that is involved in the uncoating of clathrin- coated vesicles by hsc70 in non-neuronal cells.
Western Blot: Human Brain Tissue, 1:200-1:2000
Type: Primary
Antigen: GAK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat