Anti-OIP5 Rabbit Polyclonal Antibody

Supplier: Proteintech
12142-1-AP-150UL
10091-568EA 402 USD
10091-568
Anti-OIP5 Rabbit Polyclonal Antibody
Antibodies
OIP5, also named as IS18B and CT86, is required for recruitment of CENPA to centromeres and normal chromosome segregation during mitosis. It is over expressed in the majority of primary lung and esophageal cancers. OIP5 is a promising target for developing new prognostic biomarkers and anti-cancer drugs.

Western Blot:Jurkat Cells, 1:500-1:5000

Type: Primary
Antigen: OIP5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR