Anti-RFX5 Rabbit Polyclonal Antibody

Supplier: Proteintech
12137-1-AP-150UL
10093-606EA 402 USD
10093-606
Anti-RFX5 Rabbit Polyclonal Antibody
Antibodies
Western Blot:Jurkat Cells, 1:500-1:5000

Type: Primary
Antigen: RFX5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR