Anti-NXT2 Rabbit Polyclonal Antibody

Supplier: Proteintech
12136-1-AP-150UL
10091-508EA 402 USD
10091-508
Anti-NXT2 Rabbit Polyclonal Antibody
Antibodies
IHC: Human Colon cancer, 1:20-1:200

Type: Primary
Antigen: NXT2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR