Anti-SNAI2 Rabbit Polyclonal Antibody

Supplier: Proteintech
12129-1-AP-150UL
10094-908EA 537.7 USD
10094-908
Anti-SNAI2 Rabbit Polyclonal Antibody
Antibodies
SNAI2 belongs to the Snail family of zinc finger transcription factors, which shares an evolutionarily conserved role in mesoderm formation in vertebrates and invertebrates. It tiggers epithelial-mesenchymal transitions and has a crucial role in developmental processes. Also it is a transcriptional reperssor in mediating RAF-induced expression of TJ protein, occludin(OCLN) and subsequent oncogenic transformation of epithelial cells

Western Blot: Mouse pancreas Tissue, 1:200-1:1000; IHC: Human Kidney Tissue, 1:20-1:200

Type: Primary
Antigen: SNAI2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR