Anti-PHF21A Rabbit Polyclonal Antibody

Supplier: Proteintech
12127-1-AP-150UL
10092-194EA 459.7 USD
10092-194
Anti-PHF21A Rabbit Polyclonal Antibody
Antibodies
Western Blot: HEK-293 Cells, 1:200-1:2000

Type: Primary
Antigen: PHF21A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR