Anti-WSB2 Rabbit Polyclonal Antibody

Supplier: Proteintech
12124-2-AP-150UL
10097-148EA 459.7 USD
10097-148
Anti-WSB2 Rabbit Polyclonal Antibody
Antibodies
WSB2, also named SBA2, encodes WD repeat and SOCS box-containing protein 2. WSB2 may be a substrate-recognition component of an E3 ubiquitin ligase complex which mediates the ubiquitination and subsequent proteasomal degradation of target proteins. WSB2 is a novel protein supposed to be involved in male- and female-specific gonadal development, and also a regulator of IL-21 receptor expression and IL21-induced signal transduction. WSB2 might be a promising novel biomarker of cell differentiation in colorectal cancer and its biological functions need further studies.

Western Blot: Mouse Kidney Tissue, 1:500-1:5000; IHC: Human prostate cancer, 1:20-1:200

Type: Primary
Antigen: WSB2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR