Anti-RAD23B Rabbit Polyclonal Antibody

Supplier: Proteintech
12121-1-AP-150UL
10093-284EA 459.7 USD
10093-284
Anti-RAD23B Rabbit Polyclonal Antibody
Antibodies
RAD23B, also named as p58, hHR23B and HR23B, belongs to the RAD23 family. It plays a central role both in proteosomal degradation of misfolded proteins and DNA repair. RAD23B is a central component of a complex required to couple deglycosylation and proteasome-mediated degradation of misfolded proteins in the endoplasmic reticulun that are retrotranslocated in the cytosol. Involved in DNA excision repair by stabilizing XPC protein. It may play a part in DNA damage recognition and/or in altering chromatin structure to allow access by damage-processing enzymes. RAD23B has two subunit: Heterodimer of a 125 kDa subunit (p125) and of a 58 kDa subunit (p58).

Western Blot:Jurkat Cells, 1:200-1:2000; IHC: Human breast cancer, 1:20-1:200

Type: Primary
Antigen: RAD23B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR