Anti-LRDD Rabbit Polyclonal Antibody

Supplier: Proteintech
12119-1-AP-150UL
10089-784EA 459.7 USD
10089-784
Anti-LRDD Rabbit Polyclonal Antibody
Antibodies
Western Blot:L02 Cells, 1:200-1:1000; IHC: Human pancreas Tissue, 1:20-1:200; IF: Hela Cells, 1:10-1:100; IP:L02 Cells, 1:200-1:2000

Type: Primary
Antigen: LRDD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR