Anti-PADI2 Rabbit Polyclonal Antibody

Supplier: Proteintech
12110-1-AP-150UL
10091-732EA 614.88 USD
10091-732
Anti-PADI2 Rabbit Polyclonal Antibody
Antibodies
PADI2, also named as KIAA0994, PDI2, PAD-H19 and PAD2(Peptidylarginine deiminase II ), belongs to the protein arginine deiminase family. It catalyzes the deimination of arginine residues of proteins. PADI2 may play a regulatory role in the expression of lactation related genes via histone citrullination during diestrus . PADI2 has two isoforms with MW 75 kDa and 49 kDa.

Western Blot:BxPC-3 Cells, 1:500-1:5000; IHC: Human gliomas Tissue, 1:20-1:200; IP: Mouse Brain Tissue, 1:200-1:1000; IF:BxPC-3 Cells, 1:10-1:100

Type: Primary
Antigen: PADI2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR