Anti-PTMS Rabbit Polyclonal Antibody

Supplier: Proteintech
12106-1-AP-150UL
10093-042EA 402 USD
10093-042
Anti-PTMS Rabbit Polyclonal Antibody
Antibodies
Western Blot:U-937 Cells, 1:200-1:1000

Type: Primary
Antigen: PTMS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR