Anti-UBLCP1 Rabbit Polyclonal Antibody

Supplier: Proteintech
12099-1-AP-150UL
10096-678EA 402 USD
10096-678
Anti-UBLCP1 Rabbit Polyclonal Antibody
Antibodies
Western Blot:K-562 Cells, 1:500-1:5000; IHC: Human ovary tumor Tissue, 1:20-1:200; IF: Hela Cells, 1:20-1:200; IP:K-562 Cells, 1:200-1:2000

Type: Primary
Antigen: UBLCP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR