Anti-DNAJC19 Rabbit Polyclonal Antibody

Supplier: Proteintech
12096-1-AP-150UL
10086-006EA 402 USD
10086-006
Anti-DNAJC19 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Human Brain Tissue, 1:1000-1:10000; IF: HepG2 Cells, 1:10-1:100

Type: Primary
Antigen: DNAJC19
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR