Anti-FKBP7 Rabbit Polyclonal Antibody

Supplier: Proteintech
12092-1-AP-150UL
10087-042EA 402 USD
10087-042
Anti-FKBP7 Rabbit Polyclonal Antibody
Antibodies
FKBP7(FK506-binding protein 7) is also named as FKBP23 and belongs to immunophilin class of proteins family. This protein contains an N-terminal hydrophobic signal sequence, a PPIase motif, 2 EF-hand domains, and 2 putative N-glycosylation sites. The calculated molecular mass is about 25 kD, but becomes 33 kD following cleavage of the signal sequence. It could bind to BiP in ER in a process regulated by the concentration of Ca2+. It has 3 isoforms produced by alternative splicing.

Western Blot: Human lung Tissue, 1:500-1:5000; IHC: Human lung cancer Tissue, 1:20-1:200

Type: Primary
Antigen: FKBP7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR