Anti-REEP6 Rabbit Polyclonal Antibody

Supplier: Proteintech
12088-1-AP-150UL
10093-504EA 614.88 USD
10093-504
Anti-REEP6 Rabbit Polyclonal Antibody
Antibodies
Western Blot:BxPC-3 Cells, 1:500-1:5000; IHC: Human pancreas cancer Tissue, 1:20-1:200; IF: Mouse olfactory epithelium Tissue, N/A

Type: Primary
Antigen: REEP6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR