Anti-HMG20A Rabbit Polyclonal Antibody

Supplier: Proteintech
12085-2-AP-150UL
10088-328EA 402 USD
10088-328
Anti-HMG20A Rabbit Polyclonal Antibody
Antibodies
The HMG (high mobility group) proteins are nonhistone chromosomal proteins that is present in almost all eukaryotic cells, and it functions to stabilize nucleosome formation and acts as a transcription-factor-like protein that regulates the expression of several genes. HMG20A has a role in the initiation of neuronal differentiation by activating REST-responsive genes, and it also nvolves in the recruitment of the histone methyltransferase MLL and consequent increased methylation of histone H3 lysine 4. HMG20A (for inhibitor of BRAF35) overcomes the repressive effects of the neuronal silencer REST and activates REST-responsive genes through the modulation of histone methylation

Western Blot:Jurkat Cells, 1:500-1:5000; IHC: Human pancreas cancer, 1:20-1:200; IF: HepG2, 1:10-1:100

Type: Primary
Antigen: HMG20A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR