Anti-VTCN1 Rabbit Polyclonal Antibody

Supplier: Proteintech
12080-1-AP-150UL
10097-050EA 402 USD
10097-050
Anti-VTCN1 Rabbit Polyclonal Antibody
Antibodies
VTCN1 also named B7H4, B7X, or B7S1 is a 282 amino acid protein, which contains 2 immunoglobulin-like domains and belongs to the immunoglobulin superfamily. VTCN1 negatively regulates T-cell mediated immune response by inhibiting T-cell activation, proliferation, cytokine production and development of cytotoxicity. VTCN1 is a single-pass type I membrane protein, which is over-expressed in breast, ovarian, endometrial, renal cell and non-small-cell lung cancers. The molecular weight of VTCN1 is 31kDa, but mature VTCN1 is 50 to 80kDa glycosylated molecule with a 28kDa protein core.

Western Blot: Mouse Liver Tissue, 1:200-1:2000

Type: Primary
Antigen: VTCN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR