Anti-TRA2A Rabbit Polyclonal Antibody

Supplier: Proteintech
12079-1-AP-150UL
10096-248EA 420.09 USD
10096-248
Anti-TRA2A Rabbit Polyclonal Antibody
Antibodies
Western Blot:A549 Cells, 1:1000-1:10000

Type: Primary
Antigen: TRA2A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR