Anti-SF3A3 Rabbit Polyclonal Antibody

Supplier: Proteintech
12070-1-AP-150UL
10094-392EA 459.7 USD
10094-392
Anti-SF3A3 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Raji Cells, 1:200-1:2000; IHC: Human lung cancer Tissue, 1:20-1:200; IF: Hela Cells, 1:10-1:100; IP: Raji Cells, 1:200-1:2000

Type: Primary
Antigen: SF3A3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR