Anti-BRF2 Rabbit Polyclonal Antibody

Supplier: Proteintech
12056-1-AP-150UL
10083-682EA 402 USD
10083-682
Anti-BRF2 Rabbit Polyclonal Antibody
Antibodies
Human cells contain two types of RNA polymerase III transcription factor(TFIIIB), BRF1 and BRF2. BRF2,also named as BRFU, contains one TFIIB-type zinc finger. It is a general activator of RNA polymerase III transcription. It is a factor exclusively required for RNA polymerase III transcription of genes with promoter elements upstream of the initiation sites. BRF2 is recruited to type 3 promoters such as the human U6 snRNA promoter. It differs from BRF1-TFIIIB in that it contains the TFIIB-related factor BRF2 iinstead of BRF1 and its three components do not form a stable complex. This antibody is a rabbit polyclonal antibody raised against residues near the C terminus of human BRF2. It recognizes a 50-60kd band in WB analysis.

Western Blot: HT-1080 Cells, 1:200-1:2000

Type: Primary
Antigen: BRF2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR