Anti-PCSK7 Rabbit Polyclonal Antibody

Supplier: Proteintech
12044-1-AP-150UL
10091-950EA 402 USD
10091-950
Anti-PCSK7 Rabbit Polyclonal Antibody
Antibodies
PCSK7(Proprotein convertase subtilisin/kexin type 7) is also named as LPC, PC7, PC8, SPC7 and belongs to the peptidase S8 family. It has ubiquitous endoprotease activity within constitutive secretory pathways and capable of cleavage at the RXXX[KR]R consensus motif. The full length protein has 4 glycosylation sites ans it can be N-glycosylated.

IHC: Human Colon cancer Tissue, 1:20-1:200; Western Blot: Mouse spleen Tissue, 1:200-1:1000

Type: Primary
Antigen: PCSK7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR