Anti-CHI3L1 Rabbit Polyclonal Antibody

Supplier: Proteintech
12036-1-AP-150UL
10084-702EA 402 USD
10084-702
Anti-CHI3L1 Rabbit Polyclonal Antibody
Antibodies
IHC: Human ovary tumor, 1:20-1:200

Type: Primary
Antigen: CHI3L1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR