Anti-SPATA7 Rabbit Polyclonal Antibody

Supplier: Proteintech
12020-1-AP-150UL
10095-080EA 402 USD
10095-080
Anti-SPATA7 Rabbit Polyclonal Antibody
Antibodies
SPATA7, also named as HSD3, may be involved in retinal function. It is interesting to note that mutations in SPATA7 cause LCA and RP/ARRP , two overlapping but distinct human retinal diseases. SPATA7 is another LCA and Juvenile RP Gene. It is a highly conserved, vertebrate-specific protein which expressed in the mature retina. For some modification, the MW always migrate 5-8kd. The antibody can reoginze all the 3 isoforms of SPATA7.

Western Blot: Mouse Testis Tissue, 1:200-1:2000; IHC: Human Testis Tissue, 1:20-1:200; IP: Mouse Testis Tissue, 1:200-1:1000

Type: Primary
Antigen: SPATA7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR