Anti-CHST10 Rabbit Polyclonal Antibody

Supplier: Proteintech
12013-1-AP-150UL
10084-758EA 459.7 USD
10084-758
Anti-CHST10 Rabbit Polyclonal Antibody
Antibodies
Western Blot:BxPC-3 Cells, 1:500-1:5000; IHC: Human gliomas, 1:20-1:200

Type: Primary
Antigen: CHST10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR