Anti-UBE2G1 Rabbit Polyclonal Antibody

Supplier: Proteintech
12012-1-AP-150UL
10096-636EA 402 USD
10096-636
Anti-UBE2G1 Rabbit Polyclonal Antibody
Antibodies
Western Blot: HeLa Cells, 1:200-1:2000; IF: HepG2, 1:10-1:100

Type: Primary
Antigen: UBE2G1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR