Anti-VPS45 Rabbit Polyclonal Antibody

Supplier: Proteintech
12006-1-AP-150UL
10081-342EA 459.7 USD
10081-342
Anti-VPS45 Rabbit Polyclonal Antibody
Antibodies
The Sec1/Munc18 (SM) family of proteins regulate intracellular trafficking through interactions with individual SNARE (soluble N-ethylmeleimide-sensitive factor attachment protein receptor) proteins and assembled SNARE complexes. The SM protein VPS45 (Vacuolar protein sorting-associated protein 45) has a calculated molecular mass of 65 kDa and shares 38% identity with yeast Vps45 which is required for the formation of yeast endosomal SNARE complexes and is thus essential for traffic through the endosomal system. VPS45 may play a role in vesicle-mediated protein trafficking from the Golgi stack through the trans-Golgi network.

IHC: Human gliomas Tissue, 1:20-1:200; IF: HepG2 Cells, 1:20-1:200

Type: Primary
Antigen: VPS45
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR