Anti-GALC Rabbit Polyclonal Antibody

Supplier: Proteintech
11991-1-AP-150UL
10087-326EA 402 USD
10087-326
Anti-GALC Rabbit Polyclonal Antibody
Antibodies
The GALC antibody targets the liposomal enzyme Galactosylceramidase (GALC), which belongs to the glycosyl hydrolase 59 family. It hydrolyzes the galactose ester bonds of galactosylceramide, galactosylsphingosine, lactosylceramide, and monogalactosyldiglyceride. It is primarily found in the brain and kidneys where galactolipids are hydrolyzed. Deficiencies of GALC are primarily associated with the autosomal recessive Krabbe’s disease. This disease is characterized by developmental delay caused by apoptosis of myelin-forming cells. GALC is responsible for hydrolyzing galactosylceramide, a cerebroside that is an important component of myelin. A deficiency in GALC causes loss of myelin to nerve cells, resulting in delayed nerve transmissions. Krabbe’s disease has varying degrees of severity due to a large number of different genetic mutations in the gene. The GALC antibody can be used to detect the deletions in the GALC gene and functions of the enzyme.Normal GALC mRNA encodes the 80 kDa precursor, which is processed into 50 and 30 kDa subunits.

Western Blot: Mouse Brain Tissue, 1:500-1:5000; IHC: Human gliomas Tissue, 1:20-1:200

Type: Primary
Antigen: Galc
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR