Anti-ROBO3 Rabbit Polyclonal Antibody

11982-1-AP
10093-836EA 365.28 USD
10093-836
Anti-ROBO3 Rabbit Polyclonal Antibody
Western Blot: Mouse Stomach Tissue, 1:200-1:2000; IHC: Human gliomas Tissue, 1:20-1:200

Type: Primary
Antigen: ROBO3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR