Anti-SNF8 Rabbit Polyclonal Antibody

11980-1-AP
10094-932EA 340.25 USD
10094-932
Anti-SNF8 Rabbit Polyclonal Antibody
Type: Primary
Antigen: SNF8
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR