Anti-DNAJC2/MPP11 Rabbit Polyclonal Antibody

Supplier: Proteintech
11971-1-AP-150UL
10086-008EA 402 USD
10086-008
Anti-DNAJC2/MPP11 Rabbit Polyclonal Antibody
Antibodies
Western Blot:K-562 Cells, 1:500-1:5000; IF: HepG2 Cells, 1:10-1:100

Type: Primary
Antigen: DNAJC2/MPP11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR