Anti-BRIT1 Rabbit Polyclonal Antibody

Supplier: Proteintech
11962-1-AP-150UL
10083-684EA 402 USD
10083-684
Anti-BRIT1 Rabbit Polyclonal Antibody
Antibodies
Primary microcdphaly refers to the clinical finding of a head circumference less than 3 standard deviations(SD) below the age- and sex-related mean, present at birth. MCPH1, the fisrt gene identified as causative for primary microcephaly, encodes a multifunctional protein that notably is linked to DNA damage checkpoint, DNA repair by homologous recombination and DNA transcription. MCPH1 is associated with premature chromosome condensation in particular. There are two mainly isoforms encoded by MCPH1 gene, isoform1(~100kd) and isoform2(~66kd)

Western Blot: HeLa Cells, 1:200-1:2000; IHC: Human Prostate Cancer Tissue, 1:20-1:200

Type: Primary
Antigen: BRIT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR