Anti-PCBD2 Rabbit Polyclonal Antibody

Supplier: Proteintech
11958-1-AP-150UL
10091-908EA 402 USD
10091-908
Anti-PCBD2 Rabbit Polyclonal Antibody
Antibodies
PCBD2(Pterin-4-alpha-carbinolamine dehydratase 2) is also named DCOH2, DCOHM and belongs to the pterin-4-alpha-carbinolamine dehydratase family. It is involved in tetrahydrobiopterin biosynthesis and it seems to both prevent the formation of 7-pterins and accelerate the formation of quinonoid-BH2. PCBD2 also regulates the dimerization of homeodomain protein HNF-1-alpha and enhances its transcriptional activity.

Western Blot: Human Skeletal Muscle Tissue, 1:200-1:2000

Type: Primary
Antigen: PCBD2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR