SUB1 gene encodes activated RNA polymerase II transcriptional coactivator p15, also termed as positive cofactor 4 (PC4) or p14. SUB1 is a sort of general coactivator that functions in assembling and stablization of transcription complex. It has DNA binding activity and is localized in the nucleus, mulitple modification sites of phosphorylation or acetylation may turn it appear slightly shifted molecular weight. Catalog#11956-1-AP is a rabbit polyclonal antibody raised against full-length human SUB1. This antibody recognize 14-16 kDa (P14 or P15), 20 kDa(phospho form) and 30 kDa (Dimer form).
Western Blot: HeLa Cells, 1:500-1:5000; IHC: Human gliomas, 1:20-1:200
Type: Primary
Antigen: SUB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse