Anti-HELLS Rabbit Polyclonal Antibody

Supplier: Proteintech
11955-1-AP-150UL
10088-152EA 402 USD
10088-152
Anti-HELLS Rabbit Polyclonal Antibody
Antibodies
Lymphoid-specific helicase(HELLS) ia a member of the SNF2 family of chromatin remodeling proteins that is required for development and survival. It's required for de novo or maintenance DNA methylation, which mediated the silencing of the imprinted CDKN1C gene. Also it involved in formation and organization of heterochromatin that regulate transcription and mitosis.

Western Blot:K-562 Cells, 1:1000-1:10000; IHC: Human Prostate Cancer Tissue, 1:20-1:200; IF: HepG2 Cells, 1:10-1:100

Type: Primary
Antigen: HELLS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR