Anti-ASB9 Rabbit Polyclonal Antibody

Supplier: Proteintech
11952-1-AP-150UL
10083-182EA 402 USD
10083-182
Anti-ASB9 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Human Testis Tissue, 1:200-1:2000

Type: Primary
Antigen: ASB9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR