Anti-RAB5A Rabbit Polyclonal Antibody

Supplier: Proteintech
11947-1-AP-150UL
10093-214EA 614.88 USD
10093-214
Anti-RAB5A Rabbit Polyclonal Antibody
Antibodies
Western Blot: Mouse Brain Tissue, 1:200-1:2000; IHC: Human gliomas Tissue, 1:20-1:200

Type: Primary
Antigen: RAB5A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR