Anti-STEAP4 Rabbit Polyclonal Antibody

Supplier: Proteintech
11944-1-AP-150UL
10095-294EA 402 USD
10095-294
Anti-STEAP4 Rabbit Polyclonal Antibody
Antibodies
STEAP4 (also called STAMP2), is a member of the STEAP family. Structurally, It is a 459 amino acid protein characterized by a molecular topology of six transmembrane domains. The cytoplasmic N-terminal domain of STEAP4 shows structural similarity with bacterial and archaeal FNO oxidoreductase and STEAP4 exhibits a strong iron reductase activity. Studies in mice and human suggest that STEAP4 maybe involved in adipocyte development and metabolism, and may contribute to the normal biology of the prostate cell, as well as prostate cancer progression.

Western Blot:3T3-L1 Cells, 1:500-1:5000; IHC: Human Prostate Cancer Tissue, 1:20-1:200; IP:A549 Cells, 1:500-1:5000

Type: Primary
Antigen: STEAP4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR