Anti-SGCB Rabbit Polyclonal Antibody

Supplier: Proteintech
11936-1-AP-150UL
10094-486EA 524.48 USD
10094-486
Anti-SGCB Rabbit Polyclonal Antibody
Antibodies
SGCB is a 43-kD component of the dystrophin-glycoprotein complex.

Western Blot: MCF7 Cells, 1:200-1:2000

Type: Primary
Antigen: SGCB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR