Anti-ARL15 Rabbit Polyclonal Antibody

Supplier: Proteintech
11934-1-AP-150UL
10083-120EA 599.6 USD
10083-120
Anti-ARL15 Rabbit Polyclonal Antibody
Antibodies
ARL15(ADP-ribosylation factor-like protein 15) is also named as ARFRP2(ADP-ribosylation factor-related protein 2) and belongs to the small GTPase superfamily and Arf familiy.ADP-ribosylation factor-related protein (ARP) is a membrane-associated GTPase with remote similarity to the family of ADP-ribosylation factors (ARF). Arl15 is present in chordates but not lower eukaryotes. Arl15 lacks a site for Nmyristoylation, but the N-terminal region contains several well-conserved cysteines, suggesting that it may bepalmitoylated.

Western Blot: Mouse Thymus Tissue, 1:200-1:2000; IHC: Human Lymphoma Tissue, 1:20-1:200

Type: Primary
Antigen: ARL15
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR