Anti-PRAP1 Rabbit Polyclonal Antibody

Supplier: Proteintech
11932-1-AP-150UL
10092-746EA 402 USD
10092-746
Anti-PRAP1 Rabbit Polyclonal Antibody
Antibodies
Western Blot:L02 Cells, 1:500-1:5000; IHC: Human Liver Tissue, 1:20-1:200

Type: Primary
Antigen: PRAP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR