Anti-ERGIC2 Rabbit Polyclonal Antibody

Supplier: Proteintech
11927-1-AP-150UL
10086-622EA 402 USD
10086-622
Anti-ERGIC2 Rabbit Polyclonal Antibody
Antibodies
ERGIC2, also named as Endoplasmic reticulum-Golgi intermediate compartment protein 2 or PTX1, is a 377 amino acid protein, which belongs to the ERGIC family. ERGIC2 is ubiquitously expressed in many tissues. ERGIC2 localizes in the endoplasmic reticulum-Golgi intermediate compartment membrane and may play a role in transport between endoplasmic reticulum and Golgi. ERGIC2 is involved in protein transportation, and participated in the elongation factor 1 alpha regulated mechanisms.

Western Blot: HepG2 Cells, 1:500-1:5000; IHC: Human Prostate Cancer Tissue, 1:20-1:200; IP: HepG2 Cells, 1:500-1:5000

Type: Primary
Antigen: ERGIC2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR