Anti-PLRG1 Rabbit Polyclonal Antibody

Supplier: Proteintech
11914-1-AP-150UL
10092-488EA 402 USD
10092-488
Anti-PLRG1 Rabbit Polyclonal Antibody
Antibodies
PLRG1, identifed as a component of the CDC5L complex, is one spliceosomal protein highly conserved across species. It has been shown to be essential for pre-mRNA splicing in yeast and humans.

Western Blot:K-562 Cells, 1:500-1:5000; IP: HEK-293 Cells, 1:500-1:5000

Type: Primary
Antigen: PLRG1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR