Anti-SCEL Rabbit Polyclonal Antibody

Supplier: Proteintech
11906-1-AP-150UL
10094-174EA 402 USD
10094-174
Anti-SCEL Rabbit Polyclonal Antibody
Antibodies
Western Blot:A431 Cells, 1:200-1:2000; IHC: Human cervical cancer Tissue, 1:20-1:200; IF: HepG2 Cells, 1:10-1:100; FC: HeLa Cells, N/A

Type: Primary
Antigen: SCEL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR