Anti-DGKE Rabbit Polyclonal Antibody

Supplier: Proteintech
11900-1-AP-150UL
10085-794EA 402 USD
10085-794
Anti-DGKE Rabbit Polyclonal Antibody
Antibodies
DGKE(Diacylglycerol kinase epsilon) is also named as DAGK5 and belongs to the eukaryotic diacylglycerol kinase family.It may be involved mainly in the regeneration of phosphatidylinositol (PI) from diacylglycerol in the PI-cycle during cell signal transduction.When expressed in mammalian cells, DGK-epsilon showed specificity for arachidonyl-containing diacylglycerol.

Western Blot: HEK-293 Cells, 1:500-1:5000; IHC: Human Prostate Cancer Tissue, 1:20-1:200

Type: Primary
Antigen: DGKE
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR