Anti-TERF1 Rabbit Polyclonal Antibody

Supplier: Proteintech
11899-1-AP-150UL
10095-786EA 459.7 USD
10095-786
Anti-TERF1 Rabbit Polyclonal Antibody
Antibodies
Telomeres are nucleoprotein complexes located at chromosome ends that consist of tandem arrays of TTAGGG repeats bound to specific proteins. Telomeres protect chromosome ends from degradation and end-to-end fusions, prevent activation of DNA damage checkpoints, and modulate the maintenance of telomeric DNA by telomerase. TERF1 (telomeric repeat-binding factor 1) binds the telomeric double-stranded TTAGGG repeat and negatively regulates telomere length. It's one component of the shelterin complex, which involved in the regulation of telomere length and protection.

Western Blot: HEK-293 Cells, 1:200-1:2000; IHC: Human gliomas, 1:20-1:200

Type: Primary
Antigen: TERF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR