Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PHF19 Rabbit Polyclonal Antibody

Supplier: Proteintech
11895-1-AP
PHD FINGER PROTEIN 19(PHF19), a protein homologous to the Drosophila 'Polycomblike' (Pcl) protein, encoded by human gene PCL, which encodes 2 deduced proteins: a short form of 207 aa and a long form of 580 aa. The PCL family proteins contains a PCL homology domain, a Tudor domain, 2 PHD fingers and a C-terminal tail. It is highly expressed in thymus and heart, while 508aa form predominant in placenta, skeletal muscle and kidney and 207aa form in liver and peripheral blood leukocytes. Both of these forms can inhibit transcriptional activity. And the long form is corelated with cancer.Acts as a transcritpional repressor. The mRNA level of PHF19 is low in many normal tissue, it's elevated in cancer-derived cell lines.

Western Blot:rat Brain Tissue, 1:200-1:1000; IP: HeLa Cells, 1:200-1:2000; IF: Mouse ES Cells, 1:20-1:200

Type: Primary
Antigen: PHF19
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat




Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
150 µL 11895-1-AP 10092-190 Each (150µl) Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Symbol PHF19
Antigen Name PHD Finger Protein 19
Reactivity Rat, Mouse, Human
Conjugation Unconjugated
Clonality Polyclonal
Antibody Type Primary
Host Rabbit
Isotype IgG
Western Blot Yes
ELISA Yes
ImmunoPrecipitation Yes
ImmunoFluorescence Yes
Immunogen Recombinant Protein
Purification Antigen affinity Purification
Storage Buffer PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
Shipping Temperature Store at -20°C
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty