Anti-PSMA3 Rabbit Polyclonal Antibody

Supplier: Proteintech
11887-1-AP-150UL
10092-892EA 402 USD
10092-892
Anti-PSMA3 Rabbit Polyclonal Antibody
Antibodies
PSMA3(Proteasome subunit alpha type-3) is also named as HC8, PSC8 and belongs to the peptidase T1A family. The proteasome is a multicatalytic protease complex that catalyzes an energy-dependent, extralysosomal proteolytic pathway responsible for selective elimination of proteins with aberrant structures and naturally occurring short-lived proteins related to metabolic regulation and cell cycle progression. It has 2 isoforms produced by alternative splicing.

Western Blot: Human lung Tissue, 1:200-1:2000; IHC: Human Lymphoma Tissue, 1:20-1:200

Type: Primary
Antigen: PSMA3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR