G protein subunit gamma-T1(GNGT1) is encoded by the transducin gamma-subunit gene, which localized on huaman chromosome 7q21.3, participating in various transmembrane signaling systems. Particularly, GNGT1 gene is specific to retinal rod photoreceptors. Present polyclonal anti-GNGT1 antibody (11884-1-AP) is produced by immunizing rabbits with full-length chain of GNGT1 and dectects an 8 kDa band in mouse retinal tissue.
Western Blot: Mouse Eye Tissue, 1:200-1:1000; IF: HeLa Cells, 1:10-1:100
Type: Primary
Antigen: GNGT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat