Anti-VPS37A Rabbit Polyclonal Antibody

Supplier: Proteintech
11870-1-AP-150UL
10097-022EA 402 USD
10097-022
Anti-VPS37A Rabbit Polyclonal Antibody
Antibodies
VPS37A, also named as HCRP1, belongs to the VPS37 family. It is a regulator of vesicular trafficking process. VPS37A is a member of the endosomal sorting complex required for transport (ESCRT) system, in upper motor neuron disease. ESCRT has been shown to play a central role in intracellular trafficking, in the maturation of multivesicular bodies and the sorting of ubiquitinated membrane proteins into internal luminal vesicles.

Western Blot: Human Liver Tissue, 1:200-1:2000; IHC: Human breast cancer, 1:20-1:200

Type: Primary
Antigen: VPS37A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR