Anti-SH2D1A Rabbit Polyclonal Antibody

Supplier: Proteintech
11868-1-AP-150UL
10094-514EA 402 USD
10094-514
Anti-SH2D1A Rabbit Polyclonal Antibody
Antibodies
SH2D1A, also named as DSHP and SAP, is an inhibitor of the SLAM self-association. It acts by blocking recruitment of the SH2-domain-containing signal-transduction molecule SHP-2 to a docking site in the SLAM cytoplasmic region. SH2D1A mediates interaction between FYN and SLAMF1. Defects in SH2D1A are a cause of lymphoproliferative syndrome X-linked type 1 (XLP1).

Western Blot: Mouse Thymus Tissue, 1:500-1:5000

Type: Primary
Antigen: SH2D1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR